Order Entry
Sweden
ContactUsLinkComponent
VWR® MiniArray, Microcentrifuge
VWR® MiniArray, Microcentrifuge
Supplier:  VWR Collection
Certificates
undefined
VWR® MiniArray, Microcentrifuge
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 521-2849
  • Model:
  • MiniArray
  • Max. capacity:
  • 2 standard microscope slides
  • Max. speed:
  • 4000 min⁻¹
  • Max. RCF:
  • 1216 ×g
  • Noise level:
  • 57 dB(A)
  • W×D×H:
  • 132×155×110 mm
  • Weight:
  • 0.75
Specifications