Order Entry
Sweden
ContactUsLinkComponent
 
VWR® pHenomenal® Electrode Stand
  662-1169
undefined
VWR® pHenomenal® Electrode Stand
  662-1169

 

  • Description:
    pHenomenal electrode stand, holds up to 4 electrodes + 1 temperature sensor
  • Seq:
    18
  • Pk:
    1

 

 

Electrode arm moves smoothly in almost any direction, holding up to four electrodes plus one temperature sensor safely and securely in selected positions. Fingertip control raises, lowers, and pivots the holder. The arm articulates at three points while maintaining electrodes in a vertical position.

  • Ideal for multiple readings
  • Accepts all standard electrodes
  • Pivots 360°, extends 53 cm