HelixMark® Peristaltic Pump Tubing
HelixMark peristaltic pump tubing is designed and manufactured for use with peristaltic pumps and other applications with higher effective forces. A special production process designed to meet these requirements ensures that the products provide greater stability and capacity to withstand stresses. The range offers sizes suitable for use with all commonly used pump types. The tubing is individually packed in 7.5 m coils.
- Utilisation of platinum-cured silicone
- Production and packaging in ISO Class 8 clean rooms
- Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
- Seamless batch traceability
- Double packed
- Customer-specific solutions available outside the HelixMark standard product portfolio
All HelixMark products are manufactured in clean rooms in compliance with the requirements of the medtech and pharmaceutical industry.
Typical hardness: 60 Shore
: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177.2600 and EP 3.1.9. Certified manufacturing in compliance with ISO 13485 quality standards. Supplied with material certificate in every packaging unit.