Test tubes, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
PS, clear and PP translucent. A range of tubes with conical or round bottoms manufactured under aseptic conditions or sterilised by irradiation.
- PS tubes crystal clear
- PP shock resistant and translucent
Suitable for sampling for laboratory testing.
Ordering information: Available with or without caps.
Specifications
- Catalog No:
- 212-5159
- 212-5157
- GOSSCC21-02
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre