Centrifuge tubes closure, Nalgene®
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
PP, autoclavable.
- Designed for both Nalgene® round bottom and conical centrifuge tubes
- A tab facilitates the handling of the closure
Ordering information: 525-2937 is for 50 ml tubes with rim and round bottom, 525-2938 is for 50 ml round bottom, rimless tubes.
Specifikationer
- Catalog No:
- 525-2936
- 525-2934
- 525-2933
- 525-2932
- 525-2939
- 525-2938
- 525-2937
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement