Order Entry
Sweden
ContactUsLinkComponent
Centrifuge tubes, disposable
Centrifuge tubes, disposable
Katalog # 525-0299
Leverantör:  NOLATO HERTILA
undefined
Centrifuge tubes, disposable
Katalog # 525-0299
Leverantör:  NOLATO HERTILA
Specifikationer
  • Closure type:
    Without cap
  • Ø×L:
    17×110 mm
  • Type:
    Conical Bottom
  • Capacity:
    15 ml
  • Pk:
    100
Specifikationer
Om denna produkt
Disposable polystyrene centrifuge tubes.