Distilling bridges, Claisen
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Still heads with Liebig condenser and Claisen attachment. Socket for distillation thermometer with 55 mm total length. Liebig condenser with straight outlet. Connection for coolant water via tube connector.