Safety spectacles, uvex futura
111-8017
: UVEX
- Type:Anti-UV, anti-scratch, anti-fog, anti-static
- Colour:Brown
- Description:uvex supravision plus
- Lenses:Clear PC
- Pk:1
Classic model with excellent field of vision and an outstanding fit available in a range of sizes.
- Four-position arm adjustment with integrated locking
- Adjustable lens angle
- Padded arms for greater comfort and a better fit
- Article no. 111-8019 is ideal for slim faces
Weight: 46 g
: EN 166, EN 168, EN 170