Reaction vessels, round bottom, with flat flange
Flask, round bottom, DURAN®, borosilicate glass, with flat flange with groove for vacuum use.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Maximum operating pressure at 250 °C: 1,0 bar for 2000/4000 and 6000 ml; 0,5 bar for 10000/20000 ml.
: Important: At a maximum operating temperature of 250 °C and at permissible operating pressure, the temperature difference in the glass wall of the flat flange reaction vessels may not exceed 30 °C.