Order Entry
Sweden
ContactUsLinkComponent
Racks, microcentrifuge tube
Racks, microcentrifuge tube
Catalog # 211-2629
Leverantör:  Brand
undefined
Racks, microcentrifuge tube
Catalog # 211-2629
Leverantör:  Brand

Specifikationer

  • For tubes:
    1,5 ml
  • No. of Holes:
    20
  • Colour:
    Grey
  • W×D×H:
    210×70×37 mm
  • Pk:
    1

Specifikationer

Om denna produkt

PP, autoclavable.

  • For 20 microcentrifuge tubes
  • Numbered positions for easy sample identification