Racks for centrifuge tubes, wire
Catalog # 525-0370
Leverantör: VICTORSSON INDUSTRIER
Specifikationer
- Hole Ø:17 mm
- No. of Holes:44 (4×11)
- Pk:1
Specifikationer
Om denna produkt
Nylon coated metal, autoclavable.
Specially designed for blood samples and transport.