Order Entry
Sweden
Orders LinkContactUsLinkComponent
Transfer pipettes
Transfer pipettes
Supplier:  RATIOLAB
Certificates
Transfer pipettes
Transfer pipettes
20 Options Available Below
Product Details & Documents
PE, disposable.
  • Soft, elastic bulb
  • Unbreakable, reduced risk of injury compared to glass
  • Inert to biological fluids and most acids
  • Graduated pipettes with clearly visible embossed graduations
  • Sterile pipettes are gas-sterilised and individually packed
  • Suitable for food contact
Product Family Options
Product Information
  • Capacity
    Drop vol.
    Length
    Sterile
    Packed
    Availability
    Price
  • RATI2655181
    Graduated
    Graduated
    1,0 ml
    14 µl
    144 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1 350,00 kr
    1 350,00 kr
    1 * 500 Items
       1 350,00 kr
  • 612-5472
    Graduated
    Graduated
    1,0 ml
    14 µl
    144 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 725,00 kr
    725,00 kr
    1 * 500 Items
       725,00 kr
  • RATI2600131
    Graduated
    Graduated
    2,5 ml
    30 µl
    150 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 695,00 kr
    695,00 kr
    1 * 500 Items
       695,00 kr
  • RATI2655171
    Graduated
    Graduated
    1,0 ml
    33 µl
    150 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1 075,00 kr
    1 075,00 kr
    1 * 500 Items
       1 075,00 kr
  • 612-1776
    Graduated
    Graduated
    1,0 ml
    33 µl
    150 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 320,00 kr
    320,00 kr
    1 * 500 Items
       320,00 kr
  • 612-1740
    Graduated
    Graduated
    1,0 ml
    40 µl
    138 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 515,00 kr
    515,00 kr
    1 * 500 Items
       515,00 kr
  • 612-5471
    Graduated
    Graduated
    1,2 ml
    40 µl
    63 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 945,00 kr
    945,00 kr
    1 * 500 Items
       945,00 kr
  • 612-1746
    Graduated
    Graduated
    3,0 ml
    40 µl
    150 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 310,00 kr
    310,00 kr
    1 * 500 Items
       310,00 kr
  • Graduated
    3,0 ml
    40 µl
    155 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1 100,00 kr
    1 100,00 kr
    1 * 500 Items
       1 100,00 kr
  • RATI2655135
    Graduated
    Graduated
    2,0 ml
    45 µl
    153 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1 575,00 kr
    1 575,00 kr
    1 * 500 Items
       1 575,00 kr
  • 612-1744
    Graduated
    Graduated
    2,0 ml
    45 µl
    155 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 645,00 kr
    645,00 kr
    1 * 500 Items
       645,00 kr
  • Ungraduated
    1,0 ml
    20 µl
    104 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 400 Items
    1 * 400 Items - 885,00 kr
    885,00 kr
    1 * 400 Items
       885,00 kr
  • 612-1732
    Ungraduated
    Ungraduated
    0,2 ml
    30 µl
    130 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 830,00 kr
    830,00 kr
    1 * 500 Items
       830,00 kr
  • RATI2655151
    Ungraduated
    Ungraduated
    3,0 ml
    30 µl
    155 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1 050,00 kr
    1 050,00 kr
    1 * 500 Items
       1 050,00 kr
  • 612-1755
    Ungraduated
    Ungraduated
    3,0 ml
    33 µl
    150 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 400,00 kr
    400,00 kr
    1 * 500 Items
       400,00 kr
  • RATI2600216
    Ungraduated
    Ungraduated
    4,0 ml
    40 µl
    150 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 415,00 kr
    415,00 kr
    1 * 500 Items
       415,00 kr
  • 612-1762
    Graduated
    Graduated
    1,5 ml
    45 µl
    230 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 400 Items
    1 * 400 Items - 355,00 kr
    355,00 kr
    1 * 400 Items
       355,00 kr
  • 612-5473
    Ungraduated
    Ungraduated
    10,0 ml
    45 µl
    300 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 665,00 kr
    665,00 kr
    1 * 100 Items
       665,00 kr
  • 612-5475
    Graduated
    Graduated
    1,5 ml
    45 µl
    230 mm
    +
    Bulk
    Log in for availability
    Log in for availability
    1 * 250 Items
    1 * 250 Items - 2 025,00 kr
    2 025,00 kr
    1 * 250 Items
       2 025,00 kr
  • 612-1774
    Ungraduated
    Ungraduated
    10,0 ml
    56 µl
    170 mm
    -
    Bulk
    Log in for availability
    Log in for availability
    1 * 200 Items
    1 * 200 Items - 360,00 kr
    360,00 kr
    1 * 200 Items
       360,00 kr