PCR plates, 96-well, segmented, Axygen®
Katalog # 732-0663
Leverantör: Corning
Specifikationer
- No. of wells:96
- Skirt:Semi-skirted
- Well colour:Clear
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Pk:10
- Well volume:320 μl
Specifikationer
Om denna produkt
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
- Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
- Certified free from DNase, RNase and endotoxin for highest sample integrity
- AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 240 μl
Compatible with (in addition to standard thermal cycling): BioRad iCycler, MyiQ, iQ5, Beckman CEQ sequencers.