0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Heaters
Gas Burners
Gas lighter, Fire Boy
Gas lighter, Fire Boy
451-4260
:
FRANKEA
Gas lighter, Fire Boy
451-4260
:
FRANKEA
Description:
Gas lighter, red
Seq:
01
Pk:
1
Ergonomic design with lock and safety slide.
Electronic ignition
Flame adjustable
Refillable