Order Entry
Sweden
Orders LinkContactUsLinkComponent
Vacuum membrane filter holder
Vacuum membrane filter holder
Vacuum membrane filter holder
Certificates
Vacuum membrane filter holder
Vacuum membrane filter holder
3 Options Available Below
Product Details & Documents
Borosilicate glass, aluminium clamp.
Ideal for vacuum filtration of liquids and degassing of HPLC solvents and mobile phase. With graduation.
Filtration area: 9,6 cm².
* with stopper support base, without flask.
Delivery information: Filtration funnel with No. 8 stopper support base (514-4000); filtration unit with side arm support assembly NS 40/35, with flask (516-8921).
Product Family Options
Product Information
  • Capacity
    Material
    For filter Ø
    Availability
    Price
  • 514-4000
    Glass filter funnel with silicone stopper No. 8, base (300 ml funnel without flask)
    Glass filter funnel with silicone stopper No. 8, base (300 ml funnel without flask)
    300 ml
    Borosilicate glass
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5 125,00 kr
    5 125,00 kr
    1 * 1 Items
       5 125,00 kr
  • 516-8921
    Glass filter funnel with vacuum connection and flask (300 ml funnel with 1000 ml flask)
    Glass filter funnel with vacuum connection and flask (300 ml funnel with 1000 ml flask)
    300 ml
    Borosilicate glass
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10 125,00 kr
    10 125,00 kr
    1 * 1 Items
       10 125,00 kr
  • 516-7563
    Glass filter funnel with vacuum connection and flask (1000 ml funnel with 4000 ml flask)
    Glass filter funnel with vacuum connection and flask (1000 ml funnel with 4000 ml flask)
    1000 ml
    Borosilicate glass
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 14 675,00 kr
    14 675,00 kr
    1 * 1 Items
       14 675,00 kr
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Fritted glass support base with sidearm
    Fritted glass support base with sidearm
    516-8922
    Log in for availability
    1 * 1 Items - 6 575,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Fritted glass support base with No. 8 silicone stopper
    Fritted glass support base with No. 8 silicone stopper
    516-7567
    Log in for availability
    1 * 1 Items - 3 325,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Glass flask, 1 L for membrane filter funnel 47 mm Ø
    Glass flask, 1 L for membrane filter funnel 47 mm Ø
    516-8923
    Log in for availability
    1 * 1 Items - 3 725,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Glass flask, 4 L for membrane filter funnel 47 mm Ø
    Glass flask, 4 L for membrane filter funnel 47 mm Ø
    516-7566
    Log in for availability
    1 * 1 Items - 6 175,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Glass funnel, 1 L for membrane filter funnel 47 mm Ø
    Glass funnel, 1 L for membrane filter funnel 47 mm Ø
    516-7565
    Log in for availability
    1 * 1 Items - 3 350,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Glass funnel, 300 ml for membrane filter funnel 47 mm Ø
    Glass funnel, 300 ml for membrane filter funnel 47 mm Ø
    516-7564
    Log in for availability
    1 * 1 Items - 2 825,00 kr