Order Entry
Sweden
Orders LinkContactUsLinkComponent
MicroFunnel™ Plus Filter Funnels
MicroFunnel™ Plus Filter Funnels
MicroFunnel™ Plus Filter Funnels
Certificates
MicroFunnel™ Plus Filter Funnels
7 Options Available Below
Product Details & Documents
Ideal for monitoring purified water systems maintained at temperatures up to 90 °C (194 °F).
  • Multifunctional
  • Leakproof lid
  • Squeeze separation
  • Available in two capacities
Units function as sample container and filter funnel in one which eliminates a sample transfer step and protects critical samples from contamination.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications (not compatible with hot water applications), 0,2 and 0,45 µm Supor® (polyethersulfone) membrane to minimize binding of inhibitory substances, or Metricel® Black (modified polyethersulfone) membrane (not compatible with hot water applications) for better contrast with light colored colonies.
Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
MicroFunnel™ Plus AP disposable filter funnels allow for aseptic collection of a sample through the sample port on the lid of the filter funnel without removal of the lid. Optional filter holders are recommended for use when collection hot water samples.
Supor®: Hydrophilic polyethersulfone, white
Ordering information: Filter funnels PALL4829, PALL4807 and PALL4808 are not compatible with hot water applications; for use with ambient temperature samples only. Other disposable filter funnels are available. See MicroFunnel™ Disposable filter funnels for a broad selection of disposable filter funnels, individually bagged for general testing of aqueous solutions. See MicroFunnel™ ST Disposable filter funnels for a convenient and economical choice for sterility testing within isolators. Sentino™ Microbiology pump - designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
Specifications for Product Family
Specifications
  • Catalog No:
  • 516-9891
  • 516-1238
  • 516-9890
  • 515-0052
  • 515-0055
  • 516-1239
  • 515-0157
  • Effective filtration area:
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • Max. temperature:
  • 90 °C
  • 90 °C
  • Ambient
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • Dimensions:
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 89H×87W mm
  • 76H×61W mm
  • 89H×87W mm
  • 76H×61W mm
Specifications
Product Family Options
Product Information
  • Capacity
    Material
    Membrane
    Pore size
    Availability
    Price
  • 516-9891
    MicroFunnel™ Plus filter funnel
    MicroFunnel™ Plus filter funnel
    100 ml
    PP/PS/PE with PES-membran
    Supor® white, gridded
    0,2 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 6 900,00 kr
    6 900,00 kr
    1 * 50 Items
       6 900,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    516-9891
    Effective filtration area
    13,46 cm²
    Max. temperature
    90 °C
    Dimensions
    76H×61W mm
  • 516-1238
    MicroFunnel™ Plus filter funnel
    MicroFunnel™ Plus filter funnel
    100 ml
    PE
    Supor® white, gridded
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 7 250,00 kr
    7 250,00 kr
    1 * 50 Items
       7 250,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    516-1238
    Effective filtration area
    13,46 cm²
    Max. temperature
    90 °C
    Dimensions
    76H×61W mm
  • 516-9890
    MicroFunnel™ Plus filter funnel
    MicroFunnel™ Plus filter funnel
    100 ml
    PP/PS/PE with CE-membran
    Metricel black, gridded
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 7 000,00 kr
    7 000,00 kr
    1 * 50 Items
       7 000,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    516-9890
    Effective filtration area
    13,46 cm²
    Max. temperature
    Ambient
    Dimensions
    76H×61W mm
  • 515-0052
    MicroFunnel™ Plus filter funnel
    MicroFunnel™ Plus filter funnel
    300 ml
    PP/PS/PE with PES-membran
    Supor® white, gridded
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 3 300,00 kr
    3 300,00 kr
    1 * 20 Items
       3 300,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    515-0052
    Effective filtration area
    13,46 cm²
    Max. temperature
    90 °C
    Dimensions
    89H×87W mm
  • 515-0055
    MicroFunnel™ Plus filter funnel
    MicroFunnel™ Plus filter funnel
    100 ml
    PP/PS/PE with PES-membran
    Supor® white, gridded
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 6 825,00 kr
    6 825,00 kr
    1 * 50 Items
       6 825,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    515-0055
    Effective filtration area
    13,46 cm²
    Max. temperature
    90 °C
    Dimensions
    76H×61W mm
  • 516-1239
    MicroFunnel™ Plus filter funnel
    MicroFunnel™ Plus filter funnel
    300 ml
    PE
    Supor® white, gridded
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 3 500,00 kr
    3 500,00 kr
    1 * 20 Items
       3 500,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    516-1239
    Effective filtration area
    13,46 cm²
    Max. temperature
    90 °C
    Dimensions
    89H×87W mm
  • 515-0157
    MicroFunnel™ Plus AP filter funnel
    MicroFunnel™ Plus AP filter funnel
    100 ml
    PP/PS/PE with PES-membran
    Supor® white, gridded
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 7 725,00 kr
    7 725,00 kr
    1 * 50 Items
       7 725,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    515-0157
    Effective filtration area
    13,46 cm²
    Max. temperature
    90 °C
    Dimensions
    76H×61W mm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Autoclavable adapters for vacuum manifold when using rubber stopper
    Autoclavable adapters for vacuum manifold when using rubber stopper
    516-7706
    Log in for availability
    1 * 3 Items - 1 025,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Lid kit accessory for culturing membrane in place
    Lid kit accessory for culturing membrane in place
    515-0172
    Log in for availability
    1 * 20 Items - 190,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Funnel holder, 300 ml
    Funnel holder, 300 ml
    515-0136
    Log in for availability
    1 * 1 Items - 3 475,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Funnel holder, 100 ml
    Funnel holder, 100 ml
    515-0135
    Log in for availability
    1 * 1 Items - 2 575,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Tube, AP Sampling
    Tube, AP Sampling
    PALL4845
    Log in for availability
    1 * 50 Items - 5 825,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Second generation vacuum pump adapter for MicroFunnel™ filter funnel for alternate pump
    Second generation vacuum pump adapter for MicroFunnel™ filter funnel for alternate pump
    PALL4863
    Log in for availability
    1 * 1 Items - 645,00 kr