Order Entry
Sweden
Orders LinkContactUsLinkComponent
Funnels, analytical, DURAN®
Funnels, analytical, DURAN®
Funnels, analytical, DURAN®
Funnels, analytical, DURAN®
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rapid filtration.
Product Family Options
  • Stem length
    Stem O-Ø
    Funnel O-Ø
  • 150 mm
    9 mm
    65 mm
    1 * 1 Items
    1 * 1 Items - 420,00 kr
    420,00 kr
    1 * 1 Items
       420,00 kr
  • 150 mm
    9 mm
    80 mm
    1 * 1 Items
    1 * 1 Items - 495,00 kr
    495,00 kr
    1 * 1 Items
       495,00 kr
  • 180 mm
    9 mm
    110 mm
    1 * 1 Items
    1 * 1 Items - 815,00 kr
    815,00 kr
    1 * 1 Items
       815,00 kr