Volumetric flasks, class A, blue markings
Supplier: HIRSCHMANN
Certificates
About this item
DURAN®, borosilicate glass, with glass stopper, conformity certified (DE-M marking).
- Blue enamelled indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification (available via website: www.hirschmannlab.de)
* In addition to DIN
**Wide neck
Certifications: DIN EN ISO 1042