Erlenmeyer culture flasks with screw caps, Nalgene®
Clear PETG, with flat or baffled bottom.
- Available with either spill-proof HDPE screw cap or vented screw cap with membrane
- Packaged individually for easy storage and handling
- Reduced risk of cross-contamination
Sterile, disposable culture flasks reduce the risk of cross-contamination. Ideal for shaker and suspension cultures, and for preparing or storing media. Moulded-in graduations indicate approximate contents.
- Catalog No:
- 214-7107
- 214-7108
- 214-7109
- 214-7110
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement