Order Entry
Sweden
Orders LinkContactUsLinkComponent
Face shields, Bionic™
Face shields, Bionic™
Supplier:   HONEYWELL SAFETY
Face shields, Bionic™
Supplier:  HONEYWELL SAFETY
Certificates
Face shields, Bionic™
3 Options Available Below
Product Details & Documents
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
  • With extended protection for brow and chin
  • Locking mechanism for added safety
  • Extra soft padding with washable or replaceable sweatband
  • Various adjustments for best comfort
Weight: 355 g.
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Certifications: EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Product Family Options
Product Information
  • Type
    Material
    Availability
    Price
  • Anti-scratch, anti-fog
    PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 075,00 kr
    1 075,00 kr
    1 * 1 Items
       1 075,00 kr
  • Uncoated
    PC
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 755,00 kr
    755,00 kr
    1 * 1 Items
       755,00 kr
  • Uncoated
    Acetate
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 780,00 kr
    780,00 kr
    1 * 1 Items
       780,00 kr
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, uncoated
    Replacement visor, uncoated
    111-0740
    PC
    Log in for availability
    1 * 1 Items - 185,00 kr
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, uncoated
    Replacement visor, uncoated
    111-2160
    Acetate
    Log in for availability
    1 * 1 Items - 300,00 kr
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, anti-scratch/anti-fog
    Replacement visor, anti-scratch/anti-fog
    111-1293
    PC
    Log in for availability
    1 * 1 Items - 475,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Replacement sweatband
    Replacement sweatband
    113-8053
    Log in for availability
    1 * 1 Items - 110,00 kr