Jar with snap-on lid
Supplier: NOLATO HERTILA
Certificates
About this item
PP, translucent.
- Very good chemical and thermal resistance
- With or without white LDPE snap-lid
- Conical shape
Ideal for collecting, storing and transporting samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904