Order Entry
Sweden
ContactUsLinkComponent
Chemical resistant overalls, Spacel Plus 3000 RA/GC/PS, PE
Chemical resistant overalls, Spacel Plus 3000 RA/GC/PS, PE
Catalog # 113-8676
Supplier:  HONEYWELL SAFETY
undefined
Chemical resistant overalls, Spacel Plus 3000 RA/GC/PS, PE
Catalog # 113-8676
Supplier:  HONEYWELL SAFETY
Specifications
  • Clothing Size:
    3XL
  • Garment application:
    Chemical protection,
    Industrial
  • Colour:
    Yellow
  • Seq:
    06
  • Model:
    3000 RA GC PS
  • Pk:
    10
Specifications
About this item
Disposable, liquidtight overalls made from three-layered PE for excellent protection against liquids (chemicals, oils, hydrocarbons).
  • Double adhesive flap and welded zipper
  • Excellent abrasion, tear and puncture resistance
  • Watertight due to 'Spaseal' welding
  • Glued gloves and integrated overboots for an increased safety
  • Yellow colour for improved visibility
Thickness: 100 µm
Colour: yellow
Certifications: Type 3/5, EN 340, EN 1511, Cat. III