Order Entry
Sweden
ContactUsLinkComponent
VWR®, Beard Covers, PP
VWR®, Beard Covers, PP
  113-8254
undefined
VWR®, Beard Covers, PP
  113-8254

 

  • Colour:
    White
  • Pk:
    100

 

 

Comfortable beard cover made from non woven PP. For non critical applications.

  • Elasticated bands
  • Welded seams
  • Latex-free

Density: 12 g/m²