Order Entry
Sweden
ContactUsLinkComponent
VWR®, Burette Clamps
VWR®, Burette Clamps
Catalog # 241-0043
Supplier:  VWR Collection
undefined
VWR®, Burette Clamps
Catalog # 241-0043
Supplier:  VWR Collection

Specifications

  • For:
    Two burettes, 8 - 16 mm rod
  • Pk:
    2

Specifications

About this item

Vinyl, black. Simple model.

  • For two burettes up to 12 mm diameter
  • Easy height adjustment
  • Can be attached to stand rods using knurled screws