Bottles, narrow neck, round, PFA, with screw caps
Supplier: VITLAB
Certificates
About this item
PFA, translucent, with PFA screw cap.
- Maximum chemical resistance
- Heat resistant from −200 to +250 °C, autoclavable
- Leakproof
Bottles with high resistance to extreme temperatures, chemicals and corrosion. Inert to practically all chemicals with the exception of molten alkaline metals, fluorine compounds and complex halogen compounds at high temperatures and under pressure. Ideal for high purity liquids.