Wash bottles, narrow neck
Leverantör: MURRAY S
Certifikat
Om denna produkt
LDPE, translucent with jet.
- Good chemical resistance
- Heat resistant up to +80 °C
- Leakproof
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis.