Order Entry
Sweden
Orders LinkContactUsLinkComponent
Dropping bottles, PP, Nalgene®
Dropping bottles, PP, Nalgene®
   Sustainable Options Available
Dropping bottles, PP, Nalgene®
   Sustainable Options Available
Certificates
Dropping bottles, PP, Nalgene®
2 Options Available Below
Product Details & Documents
PPCO with non-drip closure and cap made from blue PP.
  • Autoclavable even when full with tips uncapped
  • Leakproof closure
  • Integral non-drip closure with attached cap
Ideal for sterile solutions.
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-4432
  • NALGDS2420-0250
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 125 ml
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 825,00 kr
    825,00 kr
    1 * 6 Items
       825,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-4432
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 930,00 kr
    930,00 kr
    1 * 6 Items
       930,00 kr
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALGDS2420-0250
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact