Order Entry
Sweden
Orders LinkContactUsLinkComponent
Double bosshead
Double bosshead
Thumb screw, Aluminium alloy, 16,5 mm
Catalog # 241-3168
Supplier:   BOCHEM
Double bosshead
Catalog # 241-3168
Supplier:  BOCHEM
Double bosshead
Double bosshead
Thumb screw, Aluminium alloy, 16,5 mm
Price
145,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Double bosshead made from aluminium.
Screw type: Thumb screw, M8 (Type 1).
Certifications: DIN 12895
Specifications
  • Description:
    Thumb screw
  • Material:
    Aluminium alloy
  • For rod Ø:
    16,5 mm