About this item
X-OMAT® LS is an economical, blue-tinted film for when the maximum performance of BioMax® MR, BioMax® MS, or X-OMAT® AR film is not required. X-OMAT® LS film is suitable for use with all commonly used radioisotopes and chemiluminescent probes. However, the film provides significantly less detection sensitivity than the BioMax® autoradiography films, resulting in longer exposure times.
- Economical
- Suitable for both manual and automated processing
Darkroom handling: X-OMAT® LS film should be handled under safelight conditions using a safelight lamp equipped with a Safelight Filter GBX-2 and 15 W frosted bulb (other safelight filters may result in increased background fog and/or unwanted image exposure). The safelight should be positioned at least 120 cm from the film at all times. Minimise the length of time that the film is exposed to safelight conditions.