Order Entry
Sweden
ContactUsLinkComponent
UltraClear® for histology, J.T.Baker®
UltraClear® for histology, J.T.Baker®
Supplier:  Avantor
Certificates
UltraClear® for histology, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Appearance:
    passes test
  • Aromates (as Xylene):
    max. 60 ppm
  • Solubility in Ethanol:
    passes test
  • Solubility in Xylene:
    passes test

Specification Test Results

Related Information