Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl cellulose
Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Leverantör:
Thermo Fisher Scientific
Certifikat
Expandera bilden
Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Leverantör:
Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:
9004-67-5
:
MDL-nummer:
MFCD00081763
Merck Index:
14,6040
Förvaringstemperatur:
Ambient
Relaterad information
Dokumentation
Certifikat
Sök efter LOT-nummer
SDS
msds_ALFA036718
Videor
Thermo Scientific Acroseal 25 Years
Dokumentation