Order Entry
Sweden
ContactUsLinkComponent
Anti-Tubulin Alpha Chain Mouse Monoclonal Antibody [clone: TU-01] (FITC)
Anti-Tubulin Alpha Chain Mouse Monoclonal Antibody [clone: TU-01] (FITC)
Catalog # AGRIAS214626
Supplier:  Agrisera
New Product
Anti-Tubulin Alpha Chain Mouse Monoclonal Antibody [clone: TU-01] (FITC)
Catalog # AGRIAS214626
Supplier:  Agrisera
New Product

Specifications

  • Antigen name:
    Tubulin Alpha Chain
  • Clonality:
    Monoclonal
  • Clone:
    TU-01
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    S. cerevisiae,
    Turkey,
    Pig,
    A. thaliana,
    N. tabacum,
    Human,
    Mouse
  • Format:
    Liquid at 1 mg/ml
  • UniProtKB:
    Q71U36
  • Purity:
    Conjugated with fluorescein isothiocyanate (FITC) under optimum conditions, The conjugate is purified by
    size-exclusion chromatography.
  • Molecular weight:
    54 kDa
  • Storage temperature:
    Store in the dark at 2...8 °C
  • Immunogen:
    Fraction of tubulin purified from porcine brain by two cycles of polymerization - depolymerization
  • Pk:
    1

Specifications

About this item

Recommended dilution: 1 to 4 µg/ml (Flowcytometry)

Type:
Antigen: Tubulin Alpha Chain
Clonality: Monoclonal
Clone: TU-01
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Arabidopsis thaliana, Eisenia, Nicotiana tabacum, Paramecium, Saccharomyces cerevisiae, human, mouse, pig, turkey