Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrochloric acid
Hydrochloric acid 0,5 M (0,5 N)
Hydrochloric acid 0,5 M (0,5 N)
Hydrochloric acid 0,5 M (0,5 N)
CMPLZ13411026.1L
:
CUSTOM MADE CHEMICALS LAB
New Product
:
Hydrochloric acid 0,5 M (0,5 N)
CMPLZ13411026.1L
:
CUSTOM MADE CHEMICALS LAB
New Product
:
Pk:
1 L
Seq:
5037
:
:
36.46 g/mol
:
7647-01-0
:
HCl
:
MFCD00011324