Order Entry
Sweden
ContactUsLinkComponent
Sodium hypochlorite (2.5% Cl₂) in aqueous solution Reag. Ph. Eur. 1081600
Sodium hypochlorite (2.5% Cl₂) in aqueous solution Reag. Ph. Eur. 1081600
Katalog # 87939.290
Leverantör:  VWR Chemicals
CAS-nummer:  
undefined
Sodium hypochlorite (2.5% Cl₂) in aqueous solution Reag. Ph. Eur. 1081600
Katalog # 87939.290
Leverantör:  VWR Chemicals
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifikationer

Om denna produkt

CAS-nummer:7681-52-9
 :
Formel:ClNaO
MDL-nummer:MFCD00011120
 :74.44 g/mol
Förvaringstemperatur:Ambient
EINECS:231-668-3