0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Sodium thiosulphate
Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution
Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution
FFCHFF1485000
:
FF CHEMICALS
:
Sodium thiosulphate 0.2 mol/l (0.2 N) in aqueous solution
FFCHFF1485000
:
FF CHEMICALS
:
Pk:
5 L
Seq:
5190
:
:
158.11 g/mol
:
Ambient
:
231-867-5
:
7772-98-7
:
Na₂S₂O₃
:
MFCD00003499