Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Celite® 545, filter aid, Supelco®

Supplier:   Merck
Celite® 545, filter aid, Supelco®
Supplier:  Merck
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  •  :M-Clarity™ quality level = MQ100
  • EINECS:272-489-0
Specifications for Product Family

Specification Test Results

  • Solubility:
     - 0.001 g/l (20 °C)
  • Bulk Density:
    300 kg/m3
  • pH:
    10 (100 g/l, H2O, 20 °C)
Product Family Options

Product Information