
Potassium permanganate 0.01 mol/l (0.05 N) in aqueous solution
Catalog # FFCHFF20225000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364