Coomassie® brilliant blue R-250, Sigma-Aldrich®
Catalog # 1.12553.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:825.98 g/mol
Melting Pt:174…180 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:228-060-5
CAS Number:6104-59-2
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762