Coomassie brilliant blue G-250
Catalog # ROTH9598.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:854.04 g/mol
Density:>1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:228-058-4
CAS Number:6104-58-1
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482