




Ammonia 7 M in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7664-41-7
:
Formula:NH₃
MDL Number:MFCD00011418
:17.03 g/mol
Merck Index:14,492
Density:0.779
Flash Pt:14 °C (57 °F)
Storage Temperature:Refrigerator
EINECS:231-635-3
UN:3286
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS05
:GHS09
Related Information
Frequently Bought Together