
Sudan IV, Supelco®
Catalog # 67386-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:380.45 g/mol
Merck Index:12,08538
Melting Pt:188 °C
:M-Clarity™ quality level = MQ100
EINECS:201-635-8
:GHS07
Storage Temperature:Ambient
CAS Number:85-83-6
Formula:C₂₄H₂₀N₄O
MDL Number:MFCD00003893