Specifications
- Description:EasySep™ Release Mouse APC positive selection kit, 1×10⁹ cells
- Pk:1 KIT
Specifications
About this item
Immunomagnetic positive selection of particle-free mouse cells labeled with APC-conjugated antibodies.
- Highly purified cells labelled with APC-conjugated antibodies isolated from mouse tissue in less than 40 minutes
- No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Mouse APC Positive Selection Kit is designed to isolate cells that are labeled with APC-conjugated antibodies from mouse tissue by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, APC-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.