Order Entry
Sweden
Orders LinkContactUsLinkComponent
1,6-Dibromopyrene 99%

1,6-Dibromopyrene 99%

1,6-Dibromopyrene 99%
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:27973-29-1
  • Formula:C₁₆H₈Br₂
  • MDL Number:MFCD09263746
  • Molecular Weight:360.05 g/mol
  • Boiling Pt:469.6°C
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Appearance:
    white to almost white crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 %
  • Melting point:
    228°C to 232°C
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 200,00 kr
    1 200,00 kr
    1 * 1 g
       1 200,00 kr

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 3 800,00 kr
    3 800,00 kr
    1 * 5 g
       3 800,00 kr