Tetrakis(dimethylamino)diboron
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:1630-79-1
:
Formula:C₈H₂₄B₂N₄
MDL Number:MFCD00048012
:197.93 g/mol
Boiling Pt:55…57°C (2.5mmHg)
Density:0.926
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:clear colorless liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Refractive index:1.4680 to 1.4760 (20°C, 589 nm)