Loxoprofen sodium
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:80382-23-6
:
Formula:C₁₅H₁₇NaO₃
MDL Number:MFCD00941425
:268.28 g/mol
Merck Index:13,05611
Storage Temperature:Kylskåp
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:white to light yellow crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- Water:11 to 13 %