Anti-p97/VCP Mouse Monoclonal Antibody [clone: Hs-14]
NOVUNB500-455
New Product
- Antibody type:Primary
- Antigen name:Intra Acrosomal Protein
- Antigen symbol:p97/VCP
- Clonality:Monoclonal
- Clone:Hs-14
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:7415
- Antigen synonyms:yeast Cdc48p homolog|TER ATPase|TERA|transitional endoplasmic reticulum ATPase|Valosin-containing proteinMGC148092|MGC131997|p97|IBMPFDMGC8560|valosin containing protein|15S Mg(2+)-ATPase p97 subunit
- Storage buffer:TBS (approx. pH 8,0)
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:1,0 mg/ml
- Immunogen:Freshly ejaculated human sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0,2% acetic acid and subsequently used for immunisation.
- Purification:Size Exclusion Chromatography
- Pk:0,1 mg
The p97/VCP Antibody (Hs-14) from Novus Biologicals is a mouse monoclonal antibody to p97/VCP. This antibody reacts with human, mouse. The p97/VCP Antibody (Hs-14) has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Type: Primary
Antigen: p97/VCP
Clonality: Monoclonal
Clone: Hs-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse