Order Entry
Sweden
ContactUsLinkComponent
 
Dichloroisocyanuric acid sodium salt dihydrate
Catalog # MOLE14579217-1KG
Supplier:  Molekula
CAS Number:  
Dichloroisocyanuric acid sodium salt dihydrate
Catalog # MOLE14579217-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :255.98 g/mol
Storage Temperature:Ambient
EINECS:220-767-7
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:51580-86-0
Formula:C₃Cl₂N₃NaO₃·2H₂O
MDL Number:MFCD00149404