0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vacuum Traps
Accessories for Cold Trap Bath, WCT-40
Accessories for Cold Trap Bath, WCT-40
Accessories for Cold Trap Bath, WCT-40
Katalog # WITGDH.WCT11000
Supplier:
witeg Labortechnik
New Product
Expand image
Accessories for Cold Trap Bath, WCT-40
Katalog # WITGDH.WCT11000
Supplier:
witeg Labortechnik
New Product
Specifications
Description:
Glass cold trap for WCT bath, Ø106×249H mm, pressure resistance 1,5 bar at 250 °C, borosilicate glass 3.3
Pk:
1
Specifications
About this item
Product Details
Product Details