Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyclohexane-[D12]
Cyclohexane-[D₁₂] 99.5 atom % D for NMR spectroscopy
Cyclohexane-[D₁₂] 99.5 atom % D for NMR spectroscopy
Irritant
Health
Environment
Flammable
Cyclohexane-[D₁₂] 99.5 atom % D for NMR spectroscopy
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Health
Environment
Flammable
Cyclohexane-[D₁₂] 99.5 atom % D for NMR spectroscopy
Supplier:
Thermo Fisher Scientific
About this item
Specification Test Results
Appearance:
Clear colorless Liquid
Water:
≤0.03 %
D-Enrichment:
≥99.4 %
Specification Test Results