Order Entry
Sweden
ContactUsLinkComponent
Phalloidin, iFluor™ 555
Phalloidin, iFluor™ 555
Catalog # STMC100-0853
CAS Number:  
undefined
Phalloidin, iFluor™ 555
Catalog # STMC100-0853
CAS Number:  
Specifications
  • Pk:
    300 Tests
Specifications
About this item
Fluorescent dye conjugates for detection of F-actin in cell culture experiments - 300 tests.
Phalloidin is a heptapeptide derived from the poisonous mushroom Amanita phalloides. Phalloidin binds with a high specificity to F-actin filaments in cells, preventing filament depolymerization. Fluorescently labeled phalloidin can be used in labeling, identification, and quantitation of F-actin in formaldehyde-fixed and permeabilised cells. Fluorescently labeled phalloidin has lower non-specific binding compared to actin antibodies across most species of plants and animals, which provides low background and high contrast during imaging.
CAS No: 67-68-5
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').